Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080239.1 | NP_001073708.1 | 6 - 7 | 717 | 87 | |
GenBank mRNA | BC123856.1 | - | 6 - 7 | 687 | 87 | |
BC126720.1 | - | 6 - 7 | 717 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |