Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080250.1 | NP_001073719.1 | 6 - 7 | 706 | 65 | |
GenBank mRNA | BC149327.1 | - | 5 - 6 | 659 | 65 | |
BT029907.1 | - | 6 - 7 | 706 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |