Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080911.1 | NP_001074380.1 | 5 - 6 | 1188 | 82 | |
GenBank mRNA | BT021649.1 | - | 5 - 6 | 1188 | 82 | |
BT026258.1 | - | 5 - 6 | 1187 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |