Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081513.1 | NP_001074982.1 | 7 - 8 | 733 | 60 | |
GenBank mRNA | BC133374.1 | - | 7 - 8 | 733 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |