Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081516.1 | NP_001074985.1 | 2 - 3 | 351 | 91 | |
GenBank mRNA | BC133377.1 | - | 2 - 3 | 351 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |