Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTHRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081536.2 | NP_001075005.1 | 1 - 2 | 268 | 82 | |
GenBank mRNA | BC133461.1 | - | 1 - 2 | 269 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |