Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081546.1 | NP_001075015.1 | 4 - 5 | 403 | 103 | |
GenBank mRNA | BC122626.1 | - | 4 - 5 | 403 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |