Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081594.1 | NP_001075063.1 | 2 - 3 | 648 | 64 | |
GenBank mRNA | BC133336.1 | - | 2 - 3 | 648 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |