Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082460.2 | NP_001075929.1 | 3 - 4 | 379 | 56 | |
GenBank mRNA | BC133560.1 | - | 3 - 4 | 380 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |