Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082473.1 | NP_001075942.1 | 3 - 4 | 2707 | 80 | |
GenBank mRNA | BC133540.1 | - | 3 - 4 | 2707 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |