Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083375.1 | NP_001076844.1 | 8 - 9 | 920 | 150 | |
GenBank mRNA | BC114716.1 | - | 8 - 9 | 920 | 150 | |
EF661660.1 | - | 8 - 9 | 908 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |