Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083476.2 | NP_001076945.1 | 2 - 3 | 438 | 55 | |
GenBank mRNA | BC114885.1 | - | 2 - 3 | 439 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |