Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083721.1 | NP_001077190.1 | 4 - 5 | 449 | 80 | |
GenBank mRNA | BC134568.1 | - | 4 - 5 | 449 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |