Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083725.2 | NP_001077194.1 | 4 - 5 | 1176 | 92 | |
GenBank mRNA | BC105386.1 | - | 4 - 5 | 655 | 92 | |
BC134448.1 | - | 4 - 5 | 1177 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |