Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083751.1 | NP_001077220.1 | 4 - 5 | 3411 | 84 | |
GenBank mRNA | BC134621.1 | - | 4 - 5 | 3411 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |