Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083752.1 | NP_001077221.1 | 1 - 2 | 43 | 91 | |
GenBank mRNA | BC134560.1 | - | 1 - 2 | 43 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |