Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIANP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083767.1 | NP_001077236.1 | 4 - 5 | 747 | 55 | |
GenBank mRNA | BC134609.1 | - | 4 - 5 | 747 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |