Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083770.1 | NP_001077239.1 | 1 - 2 | 348 | 67 | |
GenBank mRNA | BC134608.1 | - | 1 - 2 | 348 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |