Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083775.1 | NP_001077244.1 | 3 - 4 | 435 | 60 | |
GenBank mRNA | BC134730.1 | - | 3 - 4 | 435 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |