Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083798.1 | NP_001077267.1 | 9 - 10 | 903 | 61 | |
GenBank mRNA | BC123656.1 | - | 9 - 10 | 903 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |