Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098855.1 | NP_001092325.1 | 8 - 9 | 1672 | 107 | |
GenBank mRNA | AF032924.1 | - | 2 - 3 | 315 | 107 | |
AF094699.1 | - | 7 - 8 | 971 | 107 | ||
BC142471.1 | - | 8 - 9 | 1672 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |