Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105327.2 | NP_001098797.1 | 2 - 3 | 590 | 62 | |
GenBank mRNA | BC149394.1 | - | 2 - 3 | 592 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Inventoried Probe spans exons |