Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113240.1 | NP_001106711.1 | 1 - 2 | 45 | 66 | |
NM_174113.2 | NP_776538.1 | 1 - 2 | 45 | 66 | ||
GenBank mRNA | BC142250.1 | - | 1 - 2 | 117 | 66 | |
DY085614.1 | - | 1 - 2 | 112 | 66 | ||
X54183.1 | - | 34 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |