Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113277.1 | NP_001106748.1 | 5 - 6 | 839 | 149 | |
NM_175774.3 | NP_786968.2 | 5 - 6 | 767 | 149 | ||
GenBank mRNA | BC105499.1 | - | 5 - 6 | 768 | 149 | |
V00426.1 | - | 767 | 149 | |||
V00427.1 | - | 804 | 149 | |||
V01491.1 | - | 5 - 6 | 750 | 149 | ||
V01492.1 | - | 839 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |