Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174459.3 | NP_776884.2 | 2 - 3 | 376 | 121 | |
GenBank mRNA | AB032826.1 | - | 1 - 2 | 204 | 121 | |
BC102267.1 | - | 2 - 3 | 274 | 121 | ||
CB532844.1 | - | 2 - 3 | 376 | 121 | ||
D25220.1 | - | 2 - 3 | 247 | 121 | ||
EE223056.1 | - | 2 - 3 | 274 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |