Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174462.4 | NP_776887.1 | 1 - 2 | 68 | 81 | |
GenBank mRNA | AF250035.1 | - | 1 - 2 | 43 | 81 | |
BC123536.1 | - | 1 - 2 | 69 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |