Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174533.4 | NP_776958.2 | 1 - 2 | 221 | 109 | |
GenBank mRNA | BC120234.1 | - | 1 - 2 | 212 | 109 | |
EE238321.1 | - | 1 - 2 | 222 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |