Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174603.3 | NP_777028.2 | 6 - 7 | 975 | 106 | |
GenBank mRNA | AY033938.1 | - | 6 - 7 | 941 | 106 | |
BC133291.1 | - | 6 - 7 | 975 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |