Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175798.3 | NP_786992.1 | 5 - 6 | 765 | 83 | |
GenBank mRNA | AF101437.1 | - | 5 - 6 | 714 | 83 | |
BC126708.1 | - | 5 - 6 | 765 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |