Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175798.3 | NP_786992.1 | 7 - 8 | 936 | 139 | |
GenBank mRNA | BC126708.1 | - | 7 - 8 | 936 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |