Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175823.3 | NP_787017.1 | 1 - 2 | 728 | 64 | |
GenBank mRNA | BC134634.1 | - | 1 - 2 | 728 | 64 | |
X72802.1 | - | 530 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |