Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176619.3 | NP_788792.2 | 8 - 9 | 1053 | 87 | |
GenBank mRNA | AF020510.2 | - | 8 - 9 | 1009 | 87 | |
BC133476.1 | - | 8 - 9 | 1054 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |