Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176621.3 | NP_788794.2 | 5 - 6 | 675 | 75 | |
GenBank mRNA | AB099084.1 | - | 3 - 4 | 348 | 75 | |
AF020512.2 | - | 5 - 6 | 675 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |