Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177499.3 | NP_803465.2 | 1 - 2 | 168 | 131 | |
GenBank mRNA | BC134578.1 | - | 1 - 2 | 168 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |