Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035312.2 | NP_001030389.2 | 4 - 5 | 521 | 91 | |
GenBank mRNA | BC102130.1 | - | 4 - 5 | 490 | 91 | |
DV802255.1 | - | 4 - 5 | 522 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |