Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075628.2 | NP_001069096.2 | 18 - 19 | 2886 | 114 | |
GenBank mRNA | BC149475.1 | - | 18 - 19 | 2886 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |