Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075628.2 | NP_001069096.2 | 6 - 7 | 1268 | 103 | |
GenBank mRNA | BC149475.1 | - | 6 - 7 | 1268 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |