Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075855.3 | NP_001069323.2 | 2 - 3 | 395 | 106 | |
NR_132766.1 | 2 - 3 | 395 | 106 | |||
GenBank mRNA | BC120438.1 | - | 2 - 3 | 381 | 106 | |
DT720346.1 | - | 2 - 3 | 230 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |