Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080908.3 | NP_001074377.2 | 5 - 6 | 688 | 74 | |
GenBank mRNA | DV876521.1 | - | 2 - 3 | 211 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |