Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097998.2 | NP_001091467.1 | 1 - 2 | 73 | 104 | |
GenBank mRNA | BC140623.1 | - | 1 - 2 | 74 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |