Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098040.2 | NP_001091509.2 | 2 - 3 | 278 | 72 | |
NM_001146118.1 | NP_001139590.1 | 2 - 3 | 204 | 72 | ||
GenBank mRNA | BC140495.1 | - | 2 - 3 | 176 | 72 | |
DV927834.1 | - | 2 - 3 | 278 | 72 | ||
EE386282.1 | - | 2 - 3 | 161 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |