Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPZL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098051.1 | NP_001091520.1 | 4 - 5 | 659 | 81 | |
GenBank mRNA | BC140504.1 | - | 4 - 5 | 659 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |