Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098885.1 | NP_001092355.1 | 2 - 3 | 142 | 66 | |
GenBank mRNA | BC142254.1 | - | 2 - 3 | 142 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |