Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098912.1 | NP_001092382.1 | 13 - 14 | 1192 | 110 | |
GenBank mRNA | BC142487.1 | - | 13 - 14 | 1192 | 110 | |
BC151490.1 | - | 13 - 14 | 1206 | 110 | ||
BC151824.1 | - | 13 - 14 | 1192 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |