Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098968.2 | NP_001092438.1 | 3 - 4 | 482 | 62 | |
GenBank mRNA | BC142260.1 | - | 3 - 4 | 483 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |