Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098976.2 | NP_001092446.1 | 4 - 5 | 393 | 82 | |
GenBank mRNA | BC142118.1 | - | 4 - 5 | 394 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |