Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098976.2 | NP_001092446.1 | 7 - 8 | 873 | 81 | |
GenBank mRNA | BC142118.1 | - | 7 - 8 | 874 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |