Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098986.1 | NP_001092456.1 | 2 - 3 | 186 | 70 | |
GenBank mRNA | BC142382.1 | - | 2 - 3 | 186 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |