Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098997.2 | NP_001092467.1 | 4 - 5 | 1244 | 77 | |
GenBank mRNA | BC141987.1 | - | 4 - 5 | 1245 | 77 | |
BC149426.1 | - | 4 - 5 | 1092 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |