Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100320.1 | NP_001093790.1 | 4 - 5 | 467 | 93 | |
GenBank mRNA | BT030696.1 | - | 4 - 5 | 467 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |